Assay Details
Gene Symbol: gro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202364.1 | NP_001189293.1 | 7 - 8 | 1188 | 76 | |
NM_001202365.2 | NP_001189294.1 | 8 - 9 | 1209 | 76 | ||
NM_001300610.1 | NP_001287539.1 | 7 - 8 | 1176 | 76 | ||
NM_079790.5 | NP_524514.2 | 7 - 8 | 1121 | 76 | ||
NM_170254.3 | NP_733133.1 | 7 - 8 | 1201 | 76 | ||
NM_170255.4 | NP_733134.1 | 7 - 8 | 1182 | 76 | ||
NM_170256.2 | NP_733135.1 | 7 - 8 | 1100 | 76 | ||
NM_206575.3 | NP_996298.1 | 8 - 9 | 1215 | 76 | ||
GenBank mRNA | AF145695.1 | - | 7 - 8 | 1167 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
