Assay Details
| Gene Symbol: gro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202364.1 | NP_001189293.1 | 8 - 9 | 1247 | 71 | |
| NM_001202365.2 | NP_001189294.1 | 9 - 10 | 1268 | 71 | ||
| NM_001300610.1 | NP_001287539.1 | 8 - 9 | 1235 | 71 | ||
| NM_079790.5 | NP_524514.2 | 8 - 9 | 1180 | 71 | ||
| NM_170254.3 | NP_733133.1 | 8 - 9 | 1260 | 71 | ||
| NM_170255.4 | NP_733134.1 | 8 - 9 | 1241 | 71 | ||
| NM_170256.2 | NP_733135.1 | 8 - 9 | 1159 | 71 | ||
| NM_206575.3 | NP_996298.1 | 9 - 10 | 1274 | 71 | ||
| GenBank mRNA | AF145695.1 | - | 8 - 9 | 1226 | 71 | |
More Information
Set Membership: |
|

gro
Genomic Map