Assay Details
Gene Symbol: Rb97D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014671.2 | NP_001014671.1 | 1 - 2 | 199 | 74 | |
NM_001014672.2 | NP_001014672.1 | 1 - 2 | 199 | 74 | ||
NM_001014673.2 | NP_001014673.1 | 1 - 2 | 199 | 74 | ||
NM_001014674.2 | NP_001014674.1 | 1 - 2 | 199 | 74 | ||
NM_001014675.2 | NP_001014675.1 | 1 - 2 | 199 | 74 | ||
NM_001014676.2 | NP_001014676.1 | 1 - 2 | 199 | 74 | ||
NM_001014677.2 | NP_001014677.1 | 1 - 2 | 199 | 74 | ||
NM_079796.5 | NP_524520.1 | 1 - 2 | 199 | 74 | ||
NM_170293.3 | NP_733172.1 | 1 - 2 | 199 | 74 | ||
GenBank mRNA | BT001320.1 | - | 1 - 2 | 199 | 74 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
