Assay Details
Gene Symbol: Rb97D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014672.2 | NP_001014672.1 | 5 - 6 | 1553 | 106 | |
NM_001014676.2 | NP_001014676.1 | 5 - 6 | 1539 | 106 | ||
NM_079796.5 | NP_524520.1 | 5 - 6 | 1553 | 106 | ||
NM_170293.3 | NP_733172.1 | 5 - 6 | 1535 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
