Assay Details
Gene Symbol: mRpS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079752.3 | NP_524476.1 | 2 - 3 | 265 | 56 | |
GenBank mRNA | KX531785.1 | - | 2 - 3 | 205 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
