Assay Details
| Gene Symbol: crb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043286.2 | NP_001036751.1 | 12 - 13 | 6470 | 87 | |
| NM_001260355.2 | NP_001247284.1 | 12 - 13 | 6662 | 87 | ||
| NM_001275980.1 | NP_001262909.1 | 13 - 14 | 6512 | 87 | ||
| NM_079756.3 | NP_524480.2 | 11 - 12 | 6341 | 87 | ||
More Information
Set Membership: |
|

crb
Genomic Map