Assay Details
| Gene Symbol: crb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043286.2 | NP_001036751.1 | 6 - 7 | 3334 | 58 | |
| NM_001260355.2 | NP_001247284.1 | 6 - 7 | 3526 | 58 | ||
| NM_001275980.1 | NP_001262909.1 | 7 - 8 | 3376 | 58 | ||
| NM_079756.3 | NP_524480.2 | 5 - 6 | 3205 | 58 | ||
More Information
Set Membership: |
|

crb
Genomic Map