Assay Details
| Gene Symbol: crb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043286.2 | NP_001036751.1 | 8 - 9 | 3628 | 82 | |
| NM_001260355.2 | NP_001247284.1 | 8 - 9 | 3820 | 82 | ||
| NM_001275980.1 | NP_001262909.1 | 9 - 10 | 3670 | 82 | ||
| NM_079756.3 | NP_524480.2 | 7 - 8 | 3499 | 82 | ||
More Information
Set Membership: |
|

crb
Genomic Map