Assay Details
| Gene Symbol: Syx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079761.4 | NP_524485.1 | 1 - 2 | 1088 | 73 | |
| GenBank mRNA | AY051876.1 | - | 1 - 2 | 1045 | 73 | |
More Information
Set Membership: |
|

Syx18
Genomic Map