Assay Details
Gene Symbol: RYa-R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300623.1 | NP_001287552.1 | 5 - 6 | 1499 | 82 | |
NM_001300624.1 | NP_001287553.1 | 5 - 6 | 1499 | 82 | ||
NM_079801.4 | NP_524525.3 | 5 - 6 | 1499 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
