Assay Details
| Gene Symbol: RYa-R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300623.1 | NP_001287552.1 | 7 - 8 | 1890 | 52 | |
| NM_079801.4 | NP_524525.3 | 7 - 8 | 1820 | 52 | ||
More Information
Set Membership: |
|

RYa-R
Genomic Map