Assay Details
| Gene Symbol: mRpS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079813.3 | NP_524537.1 | 1 | 1257 | 72 | |
| GenBank mRNA | BT021953.1 | - | 1 | 1235 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mRpS22| Gene Symbol: mRpS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079813.3 | NP_524537.1 | 1 | 1257 | 72 | |
| GenBank mRNA | BT021953.1 | - | 1 | 1235 | 72 | |
Set Membership: |
|