Assay Details
Gene Symbol: Hrb98DE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079819.3 | NP_524543.1 | 2 - 3 | 651 | 79 | |
NM_170370.2 | NP_733249.1 | 2 - 3 | 655 | 79 | ||
NM_170371.2 | NP_733250.1 | 2 - 3 | 643 | 79 | ||
NM_170372.2 | NP_733251.1 | 2 - 3 | 651 | 79 | ||
NM_170373.2 | NP_733252.1 | 2 - 3 | 639 | 79 | ||
NM_170374.2 | NP_733253.1 | 2 - 3 | 639 | 79 | ||
GenBank mRNA | AY061448.1 | - | 2 - 3 | 642 | 79 | |
BT099813.1 | - | 2 - 3 | 641 | 79 | ||
M15766.1 | - | 2 - 3 | 700 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
