Assay Details
Gene Symbol: AstA-R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276113.1 | NP_001263042.1 | 3 - 4 | 1741 | 88 | |
NM_079820.3 | NP_524544.1 | 3 - 4 | 1741 | 88 | ||
GenBank mRNA | AF253526.1 | - | 3 - 4 | 1092 | 88 | |
BT003199.1 | - | 3 - 4 | 1742 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
