Assay Details
| Gene Symbol: TkR99D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170301.1 | NP_001163772.1 | 7 - 8 | 1773 | 83 | |
| NM_001276163.1 | NP_001263092.1 | 7 - 8 | 1773 | 83 | ||
| NM_079832.3 | NP_524556.2 | 7 - 8 | 1773 | 83 | ||
| GenBank mRNA | BT150106.1 | - | 7 - 8 | 1773 | 83 | |
More Information
Set Membership: |
|

TkR99D
Genomic Map