Assay Details
Gene Symbol: Sry-alpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316561.1 | NP_001303490.1 | 2 | 3093 | 102 | |
NM_079841.3 | NP_524580.1 | 1 | 1648 | 102 | ||
GenBank mRNA | AY071374.1 | - | 1 | 1650 | 102 |
Gene Symbol: Sry-beta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316522.1 | NP_001303451.1 | 2 | 3093 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
