Assay Details
Gene Symbol: sima | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276171.1 | NP_001263100.1 | 4 - 5 | 2362 | 85 | |
NM_001300670.1 | NP_001287599.1 | 10 - 11 | 3636 | 85 | ||
NM_079845.4 | NP_524584.2 | 10 - 11 | 3836 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
