Assay Details
Gene Symbol: sima | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144657.3 | NP_001138129.1 | 5 - 6 | 1624 | 94 | |
NM_001300670.1 | NP_001287599.1 | 5 - 6 | 1424 | 94 | ||
NM_079845.4 | NP_524584.2 | 5 - 6 | 1624 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
