Assay Details
Gene Symbol: sima | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276171.1 | NP_001263100.1 | 2 - 3 | 1102 | 64 | |
NM_001300670.1 | NP_001287599.1 | 8 - 9 | 2376 | 64 | ||
NM_079845.4 | NP_524584.2 | 8 - 9 | 2576 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
