Assay Details
Gene Symbol: sima | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276171.1 | NP_001263100.1 | 3 - 4 | 1600 | 71 | |
NM_001300670.1 | NP_001287599.1 | 9 - 10 | 2874 | 71 | ||
NM_079845.4 | NP_524584.2 | 9 - 10 | 3074 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
