Assay Details
| Gene Symbol: CanA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260448.2 | NP_001247377.1 | 5 - 6 | 1073 | 62 | |
| NM_001260449.2 | NP_001247378.2 | 4 - 5 | 1529 | 62 | ||
| NM_079861.4 | NP_524600.3 | 5 - 6 | 1093 | 62 | ||
| GenBank mRNA | AY128480.1 | - | 5 - 6 | 1091 | 62 | |
More Information
Set Membership: |
|

CanA1
Genomic Map