Assay Details
Gene Symbol: Med | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079871.4 | NP_524610.1 | 5 - 6 | 2163 | 59 | |
NM_170559.3 | NP_733438.1 | 4 - 5 | 1941 | 59 | ||
NM_170560.2 | NP_733439.2 | 5 - 6 | 2163 | 59 | ||
GenBank mRNA | AF027729.1 | - | 5 - 6 | 2144 | 59 | |
AF041439.1 | - | 5 - 6 | 1738 | 59 | ||
AY069496.1 | - | 5 - 6 | 2138 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
