Assay Details
Gene Symbol: Med | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079871.4 | NP_524610.1 | 8 - 9 | 2742 | 71 | |
NM_170559.3 | NP_733438.1 | 7 - 8 | 2520 | 71 | ||
NM_170560.2 | NP_733439.2 | 8 - 9 | 2742 | 71 | ||
GenBank mRNA | AF027729.1 | - | 8 - 9 | 2723 | 71 | |
AF041439.1 | - | 8 - 9 | 2317 | 71 | ||
AY069496.1 | - | 8 - 9 | 2717 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
