Assay Details
| Gene Symbol: eIF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300457.1 | NP_001287386.1 | 2 - 3 | 334 | 80 | |
| NM_079989.2 | NP_524728.2 | 2 - 3 | 367 | 80 | ||
| NM_206509.1 | NP_996231.1 | 2 - 3 | 408 | 80 | ||
| GenBank mRNA | AY113498.1 | - | 2 - 3 | 367 | 80 | |
| KX531962.1 | - | 1 - 2 | 204 | 80 | ||
More Information
Set Membership: |
|

eIF1A
Genomic Map