Assay Details
| Gene Symbol: unc-45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316496.1 | NP_001303425.1 | 2 - 3 | 1526 | 69 | |
| NM_080057.4 | NP_524796.1 | 2 - 3 | 1526 | 69 | ||
| GenBank mRNA | BT150291.1 | - | 2 - 3 | 1505 | 69 | |
More Information
Set Membership: |
|

unc-45
Genomic Map