Assay Details
Gene Symbol: Fer1HCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080134.4 | NP_524873.1 | 1 - 2 | 717 | 65 | |
NM_170479.3 | NP_733358.1 | 2 - 3 | 175 | 65 | ||
NM_170480.3 | NP_733359.1 | 2 - 3 | 267 | 65 | ||
NM_170481.3 | NP_733360.1 | 2 - 3 | 227 | 65 | ||
NM_170482.3 | NP_733361.1 | 2 - 3 | 175 | 65 | ||
GenBank mRNA | AY061831.1 | - | 1 - 2 | 668 | 65 | |
BT001419.1 | - | 2 - 3 | 137 | 65 | ||
U91524.1 | - | 1 - 2 | 395 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
