Assay Details
Gene Symbol: Fur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276025.1 | NP_001262954.1 | 10 - 11 | 4195 | 82 | |
NM_080146.3 | NP_524885.1 | 10 - 11 | 4195 | 82 | ||
GenBank mRNA | L12375.1 | - | 10 - 11 | 4170 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
