Assay Details
Gene Symbol: Fur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276024.1 | NP_001262953.1 | 6 - 7 | 1898 | 78 | |
NM_001276025.1 | NP_001262954.1 | 6 - 7 | 1898 | 78 | ||
NM_080146.3 | NP_524885.1 | 6 - 7 | 1898 | 78 | ||
NM_170654.2 | NP_733102.1 | 6 - 7 | 1898 | 78 | ||
NM_170655.2 | NP_733103.1 | 6 - 7 | 1898 | 78 | ||
NM_170656.2 | NP_733104.1 | 5 - 6 | 1833 | 78 | ||
NM_206570.2 | NP_996293.1 | 7 - 8 | 1954 | 78 | ||
NM_206571.2 | NP_996294.2 | 6 - 7 | 1898 | 78 | ||
GenBank mRNA | AY069590.1 | - | 6 - 7 | 1880 | 78 | |
L12372.1 | - | 6 - 7 | 1807 | 78 | ||
L12375.1 | - | 6 - 7 | 1873 | 78 | ||
L12376.1 | - | 5 - 6 | 1787 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
