Assay Details
Gene Symbol: Fur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276025.1 | NP_001262954.1 | 9 - 10 | 3065 | 74 | |
NM_080146.3 | NP_524885.1 | 9 - 10 | 3065 | 74 | ||
GenBank mRNA | L12375.1 | - | 9 - 10 | 3040 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
