Assay Details
| Gene Symbol: mino | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170358.2 | NP_733237.1 | 1 - 2 | 282 | 71 | |
| GenBank mRNA | BT124983.1 | - | 3 - 4 | 1141 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mino| Gene Symbol: mino | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170358.2 | NP_733237.1 | 1 - 2 | 282 | 71 | |
| GenBank mRNA | BT124983.1 | - | 3 - 4 | 1141 | 71 | |
Set Membership: |
|