Assay Details
| Gene Symbol: NSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276100.1 | NP_001263029.1 | 3 - 4 | 4116 | 70 | |
| NM_170360.2 | NP_733239.1 | 3 - 4 | 4128 | 70 | ||
| GenBank mRNA | BT025221.1 | - | 3 - 4 | 4124 | 70 | |
More Information
Set Membership: |
|

NSD
Genomic Map