Assay Details
| Gene Symbol: Axn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144654.3 | NP_001138126.1 | 9 - 10 | 2966 | 60 | |
| NM_001170302.1 | NP_001163773.1 | 6 - 7 | 1991 | 60 | ||
| NM_001276168.1 | NP_001263097.1 | 9 - 10 | 2966 | 60 | ||
| NM_001276169.1 | NP_001263098.1 | 8 - 9 | 2051 | 60 | ||
| NM_170457.3 | NP_733336.1 | 10 - 11 | 3104 | 60 | ||
| NM_170458.2 | NP_733337.1 | 8 - 9 | 2051 | 60 | ||
| NM_170459.3 | NP_733338.1 | 10 - 11 | 3104 | 60 | ||
| GenBank mRNA | AF091813.1 | - | 10 - 11 | 2374 | 60 | |
| BT133166.1 | - | 6 - 7 | 1609 | 60 | ||
More Information
Set Membership: |
|

Axn
Genomic Map