Assay Details
Gene Symbol: Fur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276025.1 | NP_001262954.1 | 12 - 13 | 4437 | 66 | |
NM_170654.2 | NP_733102.1 | 11 - 12 | 3306 | 66 | ||
GenBank mRNA | L12372.1 | - | 11 - 12 | 3215 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
