Assay Details
| Gene Symbol: Fur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276024.1 | NP_001262953.1 | 9 - 10 | 3061 | 78 | |
| NM_170654.2 | NP_733102.1 | 9 - 10 | 3061 | 78 | ||
| NM_170655.2 | NP_733103.1 | 9 - 10 | 3061 | 78 | ||
| NM_170656.2 | NP_733104.1 | 8 - 9 | 2996 | 78 | ||
| NM_206570.2 | NP_996293.1 | 10 - 11 | 3117 | 78 | ||
| NM_206571.2 | NP_996294.2 | 9 - 10 | 3061 | 78 | ||
| GenBank mRNA | AY069590.1 | - | 9 - 10 | 3043 | 78 | |
| L12372.1 | - | 9 - 10 | 2970 | 78 | ||
| L12376.1 | - | 8 - 9 | 2950 | 78 | ||
More Information
Set Membership: |
|

Fur1
Genomic Map