Assay Details
| Gene Symbol: B52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014619.3 | NP_001014619.2 | 4 - 5 | 459 | 96 | |
| NM_176491.3 | NP_788668.1 | 4 - 5 | 459 | 96 | ||
| NM_176494.4 | NP_788671.2 | 4 - 5 | 469 | 96 | ||
| GenBank mRNA | BT125783.1 | - | 4 - 5 | 456 | 96 | |
More Information
Set Membership: |
|

B52
Genomic Map