Assay Details
Gene Symbol: Hsc70-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275657.1 | NP_001262586.1 | 2 | 2182 | 93 | |
NM_079632.6 | NP_524356.1 | 2 | 2214 | 93 | ||
NM_169625.2 | NP_731987.1 | 2 | 2185 | 93 | ||
NM_169626.2 | NP_731988.1 | 2 | 2126 | 93 | ||
NM_169627.2 | NP_731989.1 | 2 | 2198 | 93 | ||
NM_176502.2 | NP_788679.1 | 2 | 2130 | 93 | ||
NM_176503.2 | NP_788680.1 | 1 | 2205 | 93 | ||
GenBank mRNA | BT023894.1 | - | 2 | 2601 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
