Assay Details
Gene Symbol: p53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170223.1 | NP_001163694.1 | 7 - 8 | 1367 | 127 | |
NM_001260323.2 | NP_001247252.2 | 3 - 4 | 596 | 127 | ||
NM_206544.2 | NP_996267.1 | 7 - 8 | 1142 | 127 | ||
NM_206545.2 | NP_996268.1 | 6 - 7 | 849 | 127 | ||
GenBank mRNA | AF192555.1 | - | 6 - 7 | 833 | 127 | |
AF224713.1 | - | 6 - 7 | 826 | 127 | ||
AF224714.1 | - | 6 - 7 | 833 | 127 | ||
AF244918.1 | - | 6 - 7 | 833 | 127 | ||
AF250918.1 | - | 6 - 7 | 833 | 127 | ||
AF263722.1 | - | 6 - 7 | 849 | 127 | ||
AY069096.1 | - | 6 - 7 | 833 | 127 | ||
DQ191318.1 | - | 7 - 8 | 1082 | 127 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
