Assay Details
| Gene Symbol: Mgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143506.4 | NP_651763.4 | 7 - 8 | 1672 | 74 | |
| GenBank mRNA | AB176553.1 | - | 6 - 7 | 1027 | 74 | |
| AY047566.1 | - | 7 - 8 | 1638 | 74 | ||
More Information
Set Membership: |
|

Mgat2
Genomic Map