Assay Details
Gene Symbol: gskt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170547.3 | NP_733426.1 | 1 | 1592 | 63 | |
GenBank mRNA | BT001338.1 | - | 1 | 1592 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
