Assay Details
Gene Symbol: MBD-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141650.3 | NP_649907.1 | 1 - 2 | 274 | 150 | |
GenBank mRNA | AF171098.1 | - | 1 - 2 | 244 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
