Assay Details
| Gene Symbol: Obp19c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134548.2 | NP_608392.1 | 1 - 2 | 519 | 110 | |
| NM_167701.2 | NP_728340.2 | 1 - 2 | 330 | 110 | ||
| GenBank mRNA | BT025013.1 | - | 1 - 2 | 519 | 110 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Recommended Assay  Drosophila melanogaster | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Obp19c
Obp19c Genomic Map
Genomic Map Back To Top
 Back To Top