Assay Details
Gene Symbol: dpy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258946.1 | NP_001245875.1 | 30 - 31 | 31762 | 115 | |
NM_001258947.3 | NP_001245876.1 | 32 - 33 | 31980 | 115 | ||
NM_001258950.1 | NP_001245879.1 | 23 - 24 | 10585 | 115 | ||
NM_001273101.1 | NP_001260030.1 | 30 - 31 | 31134 | 115 | ||
NM_001273102.1 | NP_001260031.1 | 30 - 31 | 31762 | 115 | ||
NM_001273103.1 | NP_001260032.1 | 30 - 31 | 31762 | 115 | ||
NM_001273104.1 | NP_001260033.1 | 29 - 30 | 31405 | 115 | ||
NM_001273105.1 | NP_001260034.1 | 26 - 27 | 18838 | 115 | ||
NM_001273108.1 | NP_001260037.1 | 28 - 29 | 24127 | 115 | ||
NM_001273109.1 | NP_001260038.1 | 29 - 30 | 30912 | 115 | ||
NM_001273110.1 | NP_001260039.1 | 29 - 30 | 30912 | 115 | ||
NM_001273111.1 | NP_001260040.1 | 24 - 25 | 17190 | 115 | ||
NM_001273112.1 | NP_001260041.1 | 23 - 24 | 17200 | 115 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
