Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273101.1 | NP_001260030.1 | 38 - 39 | 38172 | 122 | |
| NM_001273103.1 | NP_001260032.1 | 38 - 39 | 38800 | 122 | ||
| NM_001273104.1 | NP_001260033.1 | 37 - 38 | 38443 | 122 | ||
| NM_001273105.1 | NP_001260034.1 | 34 - 35 | 25876 | 122 | ||
| NM_001273106.1 | NP_001260035.1 | 32 - 33 | 36853 | 122 | ||
| NM_001273107.1 | NP_001260036.1 | 27 - 28 | 34924 | 122 | ||
| NM_001273108.1 | NP_001260037.1 | 36 - 37 | 31165 | 122 | ||
| NM_001273109.1 | NP_001260038.1 | 37 - 38 | 37950 | 122 | ||
| NM_001273111.1 | NP_001260040.1 | 32 - 33 | 24228 | 122 | ||
| NM_001273112.1 | NP_001260041.1 | 31 - 32 | 24238 | 122 | ||
| NM_001273113.1 | NP_001260042.1 | 27 - 28 | 34924 | 122 | ||
More Information
Set Membership: |
|

dpy
Genomic Map