Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273101.1 | NP_001260030.1 | 50 - 51 | 42976 | 118 | |
| NM_001273103.1 | NP_001260032.1 | 50 - 51 | 43604 | 118 | ||
| NM_001273104.1 | NP_001260033.1 | 49 - 50 | 43247 | 118 | ||
| NM_001273105.1 | NP_001260034.1 | 46 - 47 | 30680 | 118 | ||
| NM_001273106.1 | NP_001260035.1 | 44 - 45 | 41657 | 118 | ||
| NM_001273107.1 | NP_001260036.1 | 39 - 40 | 39728 | 118 | ||
| NM_001273108.1 | NP_001260037.1 | 48 - 49 | 35969 | 118 | ||
| NM_001273109.1 | NP_001260038.1 | 49 - 50 | 42754 | 118 | ||
| NM_001273111.1 | NP_001260040.1 | 44 - 45 | 29032 | 118 | ||
| NM_001273112.1 | NP_001260041.1 | 43 - 44 | 29042 | 118 | ||
| NM_001273113.1 | NP_001260042.1 | 39 - 40 | 39728 | 118 | ||
More Information
Set Membership: |
|

dpy
Genomic Map