Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273101.1 | NP_001260030.1 | 73 - 74 | 60105 | 148 | |
| NM_001273102.1 | NP_001260031.1 | 57 - 58 | 54016 | 148 | ||
| NM_001273103.1 | NP_001260032.1 | 73 - 74 | 60733 | 148 | ||
| NM_001273104.1 | NP_001260033.1 | 72 - 73 | 60376 | 148 | ||
| NM_001273105.1 | NP_001260034.1 | 69 - 70 | 47809 | 148 | ||
| NM_001273106.1 | NP_001260035.1 | 67 - 68 | 58786 | 148 | ||
| NM_001273107.1 | NP_001260036.1 | 62 - 63 | 56857 | 148 | ||
| NM_001273108.1 | NP_001260037.1 | 71 - 72 | 53098 | 148 | ||
| NM_001273111.1 | NP_001260040.1 | 67 - 68 | 46161 | 148 | ||
More Information
Set Membership: |
|

dpy
Genomic Map