Assay Details
| Gene Symbol: Rap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202094.2 | NP_001189023.1 | 2 | 2757 | 100 | |
| NM_001274366.1 | NP_001261295.1 | 1 | 2587 | 100 | ||
| NM_057509.5 | NP_476857.1 | 2 | 2626 | 100 | ||
| GenBank mRNA | AY121688.1 | - | 2 | 2621 | 100 | |
More Information
Set Membership: |
|

Rap1
Genomic Map