Assay Details
Gene Symbol: SmB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201851.2 | NP_001188780.1 | 2 | 169 | 66 | |
GenBank mRNA | AY061171.1 | - | 2 | 333 | 66 | |
L02919.1 | - | 2 | 101 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
