Assay Details
Gene Symbol: TpnC47D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299381.1 | NP_001286310.1 | 4 | 527 | 85 | |
NM_057620.6 | NP_476968.1 | 4 | 527 | 85 | ||
GenBank mRNA | BT030769.1 | - | 4 | 533 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
