Assay Details
Gene Symbol: Acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057767.4 | NP_477115.1 | 2 | 423 | 49 | |
GenBank mRNA | AY119158.1 | - | 2 | 424 | 49 | |
KX531884.1 | - | 1 | 367 | 49 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
