Assay Details
Gene Symbol: Jafrac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298273.1 | NP_001285202.1 | 2 | 913 | 90 | |
NM_001298274.1 | NP_001285203.1 | 2 | 942 | 90 | ||
NM_001298275.1 | NP_001285204.1 | 1 | 838 | 90 | ||
NM_058162.3 | NP_477510.1 | 2 | 872 | 90 | ||
NM_167359.2 | NP_727689.1 | 2 | 876 | 90 | ||
GenBank mRNA | AF167098.1 | - | 2 | 790 | 90 | |
AF321615.1 | - | 2 | 855 | 90 | ||
AF321616.1 | - | 2 | 868 | 90 | ||
AY070534.1 | - | 2 | 852 | 90 | ||
BT014630.1 | - | 1235 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
